Skip to Content
Merck

Skip To

RAB1197

Bovine VEGF-A ELISA

for serum, plasma and cell culture supernatants

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Please contact Customer Service for Availability
13 700,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
bovine ... VEGFA(281572)
Input:
sample type serum
sample type plasma
sample type cell culture supernatant(s)
Species reactivity:
bovine
Storage temp.:
−20°C
Shipped in:
wet ice

13 700,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

bovine

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.12 ng/mL
standard curve range: 0.123-30 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

bovine ... VEGFA(281572)

General description

This ELISA antibody pair detects Bovine VEGF-A

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB1227RAB1095RAB1871
description

for serum, plasma and cell culture supernatants

description

for serum, plasma and cell culture supernatants

description

for cell culture supernatants, plasma, and serum samples

description

for serum, plasma and cell culture supernatants

Gene Information

bovine ... VEGFA(281572)

Gene Information

equine ... VEGFA(100033839)

Gene Information

human ... VEGFB(7423)

Gene Information

ovine ... VEGFA(443103)

species reactivity

bovine

species reactivity

equine

species reactivity

human

species reactivity

sheep

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

input

sample type plasma
sample type serum sample(s)
sample type cell culture supernatant(s)

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.12 ng/mL
standard curve range: 0.123-30 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 pg/mL
standard curve range: 4.10-1000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.4 ng/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 pg/mL
standard curve range: 4.10-1000 pg/mL

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters