Skip to Content
Merck

Skip To

RAB1016

Canine MMP-8 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Please contact Customer Service for Availability
13 700,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Species reactivity:
canine
Storage temp.:
−20°C
Shipped in:
wet ice

13 700,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

canine

technique(s)

ELISA: suitable

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.07 ng/ml

shipped in

wet ice

storage temp.

−20°C

General description

This ELISA antibody pair detects Canine Matrix Metallopeptidase 8 (MMP8)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB1015RAB1017RAB1201
description

for cell culture supernatants, plasma, and serum samples

description

for cell culture supernatants, plasma, and serum samples

description

for cell culture supernatants, plasma, and serum samples

description

for serum, plasma and cell culture supernatants

species reactivity

canine

species reactivity

canine

species reactivity

canine

species reactivity

canine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.07 ng/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 16 pg/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters