Skip to Content
Merck

Skip To

RAB0996

Phosphotyrosine STAT4 / Signal Transducer and Activator of Transcription 4 ELISA Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Please contact Customer Service for Availability
13 700,00 CZK

About This Item

NACRES:
NA.84
UNSPSC Code:
41116158
Gene information:
human ... STAT4(6775)
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

13 700,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... STAT4(6775)

General description

The antibody pair provided in this kit recognizes Human Tyrosine-Phosphorylated-STAT4.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0998RAB0997RAB0994
description

-

description

-

description

-

description

-

Gene Information

human ... STAT4(6775)

Gene Information

human ... STAT6(6778)

Gene Information

human ... STAT5A(6776)

Gene Information

human ... STAT1(6772)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <10%
intra-assay cv: <12%

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Phosphotyrosine ELISA HRP-Streptavidin
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters