Skip to Content
Merck

Skip To

RAB0815

Mouse Il33 / Interleukin-33 ELISA Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Please contact Customer Service for Availability
16 900,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
mouse ... Il33(77125)
Input:
sample type serum
sample type cell culture supernatant(s)
sample type plasma
Species reactivity:
mouse
Storage temp.:
−20°C
Shipped in:
wet ice

16 900,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 2.05-500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il33(77125)

General description

The antibody pair provided in this kit recognizes mouse IL-33.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0257RAB0811RAB0812
description

-

description

for serum, plasma and cell culture supernatant

description

-

description

-

Gene Information

mouse ... Il33(77125)

Gene Information

mouse ... Il13(16163)

Gene Information

mouse ... Il2Ra(16184)

Gene Information

mouse ... Il20(58181)

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 2.05-500 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 8023-2000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 10.24-2500 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 16 pg/mL
standard curve range: 16.38-4000 pg/mL

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters