Skip to Content
Merck

Skip To

RAB0597

Feline KITLG / Kit Ligand ELISA Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Please contact Customer Service for Availability
21 600,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
feline ... KITLG(493937)
Species reactivity:
feline
Storage temp.:
−20°C
Shipped in:
wet ice

21 600,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

feline

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

feline ... KITLG(493937)

General description

The KITLG ELISA kit provides for the quantitative measurement of KITLG in Cell Culture Supernatants, Plasma and Serum.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0595RAB0578RAB0560
Gene Information

feline ... KITLG(493937)

Gene Information

feline ... IL8(493836)

Gene Information

canine ... KITLG(403507)

Gene Information

mouse ... Lgals1(16852)

species reactivity

feline

species reactivity

feline

species reactivity

canine

species reactivity

mouse

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 25 pg/mL
standard curve range: 24.58-6000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 65 pg/mL
standard curve range: 0.066-16 ng/mL

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 12 pg/mL

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters