Skip to Content
Merck

Skip To

RAB0593

Feline IL4 / Interleukin-4 ELISA Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Please contact Customer Service for Availability
13 700,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
feline ... IL4(751514)
Input:
sample type plasma
sample type serum
sample type cell culture supernatant(s)
Species reactivity:
feline
Storage temp.:
−20°C
Shipped in:
wet ice

13 700,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

feline

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.3 ng/mL
standard curve range: 0.33-80 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

feline ... IL4(751514)

General description

The antibody pair provided in this kit recognizes Feline IL-4.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters