Skip to Content
Merck
All Photos(2)

Key Documents

RAB0432

Sigma-Aldrich

Mouse Shh-N ELISA Kit

for cell and tissue lysates

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
CZK 13,700.00

CZK 13,700.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
CZK 13,700.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

CZK 13,700.00


Please contact Customer Service for Availability

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type tissue lysate
sample type cell lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 4.92-1200 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Shh(20423)

General description

The Mouse Shh-N (Sonic Hedgehog N Terminus) ELSA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Mouse Shh-N cell lysate and tissue lysate.

Immunogen

Recombinant Mouse Sonic Hedgehog (N-terminus)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuqi Li et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(16), 4326-4338 (2014-06-21)
Pancreatic cancer is characterized by stromal desmoplasia and perineural invasion (PNI). We sought to explore the contribution of pancreatic stellate cells (PSC) activated by paracrine Sonic Hedgehog (SHH) in pancreatic cancer PNI and progression. In this study, the expression dynamics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service