Skip to Content
Merck

Skip To

RAB0405

Mouse PLGF-2 ELISA Kit

for serum, plasma and cell culture supernatant

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Please contact Customer Service for Availability
14 000,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
mouse ... Pgf(18654)
Input:
sample type plasma
sample type serum
sample type cell culture supernatant(s)
Species reactivity:
mouse
Storage temp.:
−20°C
Shipped in:
wet ice

14 000,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 1.64-400 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Pgf(18654)

General description

The Mouse PιGF-2 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse PιGF-2 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse Placental Growth Factor (Isoform 2)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0184RAB0214RAB0493
description

for serum, plasma and cell culture supernatant

description

for serum, plasma and cell culture supernatant

description

for serum, plasma and cell culture supernatant

description

for serum, plasma, cell culture supernatant

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

Gene Information

mouse ... Pgf(18654)

Gene Information

mouse ... Fgf2(54250)

Gene Information

mouse ... Hgf(15234)

Gene Information

mouse ... Tnfrsf11b(18383)

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 1.64-400 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8 pg/mL
standard curve range: 18.8-600 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 400 pg/mL
standard curve range: 0.614-60 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 1.65-1200 pg/mL

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 5X Assay/Sample Diluent Buffer B (Item E1)
    SDS

  • ELISA 1X Assay/Sample Diluent Buffer C (Item L)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters