Skip to Content
Merck

Skip To

RAB0131

Mouse LIX / CXCL5 ELISA Kit

for serum, plasma and cell culture supernatant

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Please contact Customer Service for Availability
13 700,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
mouse ... Cxcl5(20311)
Input:
sample type cell culture supernatant(s)
sample type serum
sample type plasma
Species reactivity:
mouse
Storage temp.:
−20°C
Shipped in:
wet ice

13 700,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 35 pg/mL
standard curve range: 31.25-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Cxcl5(20311)

General description

The Mouse LIX (LPS induced C-X-C chemokine) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Mouse LIX in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse LIX

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0139RAB0043RAB0022
Gene Information

mouse ... Cxcl5(20311)

Gene Information

mouse ... Cxcl9(17329)

Gene Information

mouse ... Ccl11(20292)

Gene Information

mouse ... Axl(26362)

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 35 pg/mL
standard curve range: 31.25-2000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 2.741-2000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 1.02-100 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 1-250 pg/mL

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 5X Assay/Sample Diluent Buffer B (Item E1)
    SDS

  • ELISA 1X Assay/Sample Diluent Buffer C (Item L)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters