Skip to Content
Merck

RAB0107

Human Cardiotrophin-1 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

CT-1

Sign In to View Organizational & Contract Pricing.

Select a Size

1 KIT

CZK 14,200.00

CZK 14,200.00


Please contact Customer Service for Availability


About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... CTF1(1435)
Input:
sample type plasma
sample type cell culture supernatant(s)
sample type serum
sample type urine
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 16.46-12000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CTF1(1435)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
RAB0273RAB0048RAB0232
species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

Gene Information

human ... CTF1(1435)

Gene Information

human ... IL1B(3553)

Gene Information

human ... CCL15(6359)

Gene Information

human ... IGFBP1(3484)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 16.46-12000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.3 pg/mL
standard curve range: 0.48-100 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 4.1-1000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 2.74-2000 pg/mL

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum
sample type urine

input

sample type urine
sample type serum
sample type plasma
sample type cell culture supernatant(s)

input

sample type plasma
sample type cell culture supernatant(s)
sample type urine
sample type serum

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)
sample type urine

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

General description

The Human CT-1 (Cardiotrophin-1) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CT-1 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human Cardiotrophin-1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
RAB0107-1KT04061834959892

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service