Skip to Content
Merck
All Photos(1)

Key Documents

RAB0006

Sigma-Aldrich

Adiponutrin EIA Kit

for serum, plasma, culture supernatant and cell lysates

Synonym(s):

ADPN

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
CZK 13,000.00

CZK 13,000.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
CZK 13,000.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

CZK 13,000.00


Please contact Customer Service for Availability

species reactivity

rat, mouse, human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

input

sample type cell lysate
sample type plasma
sample type culture supernatant(s)
sample type serum

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 21 pg/mL
standard curve range: 0.1-1000 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PNPLA3(80339)

General description

The Adiponutrin Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting Adiponutrin peptide based on the principle of Competitive Enzyme Immunoassay. It targets the common sequence of human, mouse, and rat.

Immunogen

Adiponutrin, synthetic peptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service