Skip to Content
Merck

Skip To

R6016

Unwire full racks, polyketone

Holds 72 x 13-mm tubes, white

Synonym(s):

moulded rack, test tube rack, Nalgene 5970-0013, benchtop tube storage, tube holder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
CZK 974.00

About This Item

NACRES:
NB.35
UNSPSC Code:
41122804
Manufacturer/tradename:
Nalgene® 5970-0013
Feature:
Holds 72 x 13-mm tubes
Material:
polyketone
Packaging:
pack of 1 ea

CZK 974.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polyketone

feature

Holds 72 x 13-mm tubes

packaging

pack of 1 ea

manufacturer/tradename

Nalgene® 5970-0013

L × W × H

200 mm × 102 mm × 57 mm

color

white

General description

Molded of durable polyketone. Unwire test tube racks have no coating that can chip or peel and permit corrosion. Excellent replacement for wire racks; withstands most lab chemicals. Excellent thermal properties and won′t get brittle in freezers. Grid design allows tubes to be tipped from side to side for clearly marking position. These racks will not float in water baths.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Unwire is a trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
R7266R7891R8516
manufacturer/tradename

Nalgene® 5970-0013

manufacturer/tradename

Nalgene® 5970-0213

manufacturer/tradename

Nalgene® 5970-0313

manufacturer/tradename

Nalgene® 5970-0413

feature

Holds 72 x 13-mm tubes

feature

Holds 72 x 13-mm tubes

feature

Holds 72 x 13-mm tubes

feature

Holds 72 x 13-mm tubes

material

polyketone

material

polyketone

material

polyketone

material

polyketone

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

color

white

color

yellow

color

blue

color

green

L × W × H

200 mm × 102 mm × 57 mm

L × W × H

200 mm × 102 mm × 57 mm

L × W × H

200 mm × 102 mm × 57 mm

L × W × H

200 mm × 102 mm × 57 mm


Still not finding the right product?

Explore all of our products under Unwire full racks, polyketone




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library