Skip to Content
Merck

R4142

RNA Marker template set

~0.5 mg/mL in 10 mM Tris-HCl, pH 7.5, 1 mM EDTA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

concentration

~0.5 mg/mL in 10 mM Tris-HCl, pH 7.5, 1 mM EDTA

storage temp.

−20°C

Application

In vitro transcription with T7 RNA polymerase produces transcripts 100, 200, 300, 400, 600, 800 and 1,000 bases long. The transcripts can be labeled by including biotinylated, radioactive, or other labeled rNTP′s in the reaction. The markers are useful for RNase protection assays or for electrophoresis of low molecular weight RNA fragments.

Other Notes

A mixture of seven linearized plasmids for use as templates for RNA markers.


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library