Skip to Content
Merck
All Photos(1)

Key Documents

R1885

Sigma-Aldrich

Swa I from Staphylococcus warneri

buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Specificity

Recognition sequence: 5′-ATTT/AAAT-3′
Ligation and recutting results: After 2-10-fold Swa I overdigestion of 1 μg λ DNA substrate, results in 100% cutting, >95% of fragments can be ligated, and >95% recut.
Heat inactivation: Inactivated at 65 °C for 20 minutes.

Other Notes

Supplied with 10x Restriction Endonuclease Buffer SH (B3657).

Physical form

Solution in 20 mM Tris-HCl, pH 8.0, 0.1 mM EDTA, 500 mM NaCl, 10 mM 2-mercaptoethanol, 50% glycerol (v/v) at 4 °C

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Mise et al.
Gene, 33(3), 357-361 (1985-01-01)
A new restriction endonuclease, StyI, free of contaminating nuclease activities, has been isolated from Escherichia coli carrying the hsd+ miniplasmid of Salmonella typhi origin. In the presence of 10 mM Mg2+, it recognizes and cleaves a hexanucleotide sequence of 5'-C
Jaroslav Jelinek et al.
Epigenetics, 7(12), 1368-1378 (2012-10-19)
Genome wide analysis of DNA methylation provides important information in a variety of diseases, including cancer. Here, we describe a simple method, Digital Restriction Enzyme Analysis of Methylation (DREAM), based on next generation sequencing analysis of methylation-specific signatures created by
Saqib H Ansari et al.
Journal of pediatric hematology/oncology, 35(4), e153-e156 (2013-02-08)
β-thalassemia is characterized by impaired β-chain synthesis leading to ineffective erythropoiesis, severe anemia, and a need for blood transfusion. Presence of Xmn I polymorphism (-158 C-T nucleotide change) in γ-globin gene is associated with a higher fetal hemoglobin and a
Amal M El-Shehaby et al.
Scandinavian journal of clinical and laboratory investigation, 73(1), 75-81 (2012-12-04)
Left ventricular hypertrophy (LVH) is a common manifestation of cardiovascular disease and has an important prognostic value in patients with end-stage renal disease (ESRD). Vitamin D receptor (VDR) has been intensively investigated, and one of these (BsmI) already has been
Gajendradhar R Dwivedi et al.
Nucleic acids research, 41(5), 3274-3288 (2013-01-29)
Helicobacter pylori is a Gram-negative bacterium that colonizes human stomach and causes gastric inflammation. The species is naturally competent and displays remarkable diversity. The presence of a large number of restriction-modification (R-M) systems in this bacterium creates a barrier against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service