Skip to Content
Merck
All Photos(1)

Documents

PZ0337

Sigma-Aldrich

UK-432097

≥98% (HPLC)

Synonym(s):

6-(2,2-Diphenylethylamino)-9-((2R,3R,4S,5S)-5-(ethylcarbamoyl)-3,4-dihydroxytetrahydrofuran-2-yl)-N-(2-(3-(1-(pyridin-2-yl)piperidin-4-yl)ureido)ethyl)-9H-purine2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H47N11O6
CAS Number:
Molecular Weight:
777.87
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](C(NCC)=O)O[C@@H](N2C=NC(C2=N3)=C(NCC(C4=CC=CC=C4)C5=CC=CC=C5)N=C3C(NCCNC(NC(CC6)CCN6C7=CC=CC=N7)=O)=O)[C@@H]1O

InChI

1S/C40H47N11O6/c1-2-41-37(54)33-31(52)32(53)39(57-33)51-24-46-30-34(45-23-28(25-11-5-3-6-12-25)26-13-7-4-8-14-26)48-35(49-36(30)51)38(55)43-19-20-44-40(56)47-27-16-21-50(22-17-27)29-15-9-10-18-42-29/h3-15,18,24,27-28,31-33,39,52-53H,2,16-17,19-23H2,1H3,(H,41,54)(H,43,55)(H2,44,47,56)(H,45,48,49)/t31-,32+,33-,39+/m0/s1

InChI key

ZOTHAEBAWXWVID-HXEFRTELSA-N

Biochem/physiol Actions

UK-432097 is a selective adenosine receptor A2AAR full agonist with EC50 of 0.66 ± 0.19 nM. It was recently used to map the structure of the active state of the adenosine A2A receptor.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service