Skip to Content
Merck
All Photos(3)

Key Documents

PRS3819

Sigma-Aldrich

Anti-BIK antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Apoptosis inducer NBK, Anti-BCL2-Interacting killer (apoptosis-inducing)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 18 kDa

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... BIK(638)

General description

BIK (BCL2 interacting killer) is the founding member of the BH3 (BCL2 homology domain 3)-only pro-apoptotic protein family. This protein shows major localization to the ER (endoplasmic reticulum). It shares two domains with other BCL2 (B cell lymphoma) family proteins, namely BH3 domain and the trans-membrane domain. BIK is a 160-residue phosphoprotein and is highly conserved among primates. This gene is localized to human chromosome 22q13.2.

Immunogen

a 15 amino acid peptide from near the amino-terminus of human BIK.

Biochem/physiol Actions

BIK (BCL2 interacting killer) induces the release of calcium from ER (endoplasmic reticulum) to the mitochondria and mitochondrial cristae remodeling, thus, promoting apoptosis via the mitochondrial pathway. In certain types of cells, this protein also promotes non-apoptotic cell death through yet unknown mechanisms. It is critical in apoptosis induced by viral infections, cytokines and toxins. It is responsible for the selection of mature B cells in humans, and also functions as a pro-apoptotic tumor suppressor in various human tissues. Deletions in the BIK chromosomal locus or epigenetic silencing of BIK gene results in its inactivation in multiple types of cancers. BIK is up-regulated during hepatitis C virus (HCV), mediated by the viral protein NS5B (nonstructural protein 5B), and is crucial for HCV RNA replication and viral release. Studies have shown that BikDD, a constitutively active mutant of BIK, functions as a potent anti-tumor protein in preclinical pancreatic, lung and breast cancer models. Further mutation of BikDD produces BikDDA, which has a longer half-life and has potent pro-apoptotic activity in triple-negative breast cancer (TNBC).

Linkage

The action of this antibody can be blocked using blocking peptide SBP3819.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BIK, the founding member of the BH3-only family proteins: mechanisms of cell death and role in cancer and pathogenic processes.
Chinnadurai G et al
Oncogene, Suppl 1, S20-S29 (2008)
BikDDA, a mutant of Bik with longer half-life expression protein, can be a novel therapeutic gene for triple-negative breast cancer.
Jiao S et al
PLoS ONE, 9(3), e92172-e92172 (2014)
NS5B induces up-regulation of the BH3-only protein, BIK, essential for the hepatitis C virus RNA replication and viral release.
Aweya JJ et al
Virology, 474, 41-51 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service