Skip to Content
Merck

Skip To

PLA0269

Rabbit anti-HSP60 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

60 kDa chaperonin, CPN60, GroEL, HLD4, HSP-60, HSP60, HSP65, Heat shock protein 60, HuCHA60, P60 lymphocyte protein, SPG13, chaperonin 60, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), heat shock protein 65, mitochondrial matrix protein P1, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
11 600,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
3

11 600,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:200- 1:1,000, immunoprecipitation (IP): 2-5 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_002147.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... HSP60(3329)

General description

The HSPD1 (heat shock protein family D member 1) gene encodes the HSP60 (heat shock protein 60) subunit of the HSP60/HSP10 chaperonin complex. The HSPD1 gene consisting of 12 exons is localized to human chromosome 2q33.1.

Immunogen

The epitope recognized by PLA0269 maps to a region between residue 250 and 300 of human Heat Shock Protein 60 using the numbering given in entry NP_002147.2 (GeneID 3329).

Biochem/physiol Actions

The protein encoded by the HSPD1 (heat shock protein family D member 1) gene forms a double-barrel chaperonin complex with heat shock protein 10 (HSP10). This chaperonin complex facilitates folding of proteins to their native state in the mitochondrial matrix space. Missense mutation in the HSP60 gene has been associated with a rare dominant form of hereditary spastic paraplegia and a recessively inherited white matter disorder called MitCHAP60 disease.
HSP60 is also found to interact with the transcription factor interferon regulatory factor 3 (IRF3), which is involved in the induction of IFN-β (interferon- β) signaling pathway, a crucial host-protective response.

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Heat shock protein 60 (HSP60) is a member of the chaperonin family. HSP60 is a mitochondrial protein that may function as a signaling molecule in the innate immune system. It is essential for the folding and assembly of newly imported proteins in the mitochondria. [taken from NCBI Entrez Gene (GeneID: 3329)].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4501464SAB1301520SAB5700661
description

Powered by Bethyl Laboratories, Inc.

description

affinity isolated antibody

description

IgG fraction of antiserum, buffered aqueous solution

description

-

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

species reactivity

human, mouse

species reactivity

human, rat, mouse

species reactivity

mouse, human

species reactivity

human, rat, mouse

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters