Skip to Content
Merck

Skip To

PLA0266

Rabbit anti-NOP56 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

NOL5A, NOP56 ribonucleoprotein homolog, NOP56 ribonucleoprotein homolog (yeast), Nucleolar protein 5A, SCA36, nucleolar protein 5A (56kD with KKE/D repeat), nucleolar protein 5A (56kDa with KKE/D repeat), spinocerebellar ataxia 36

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
12 300,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
-

12 300,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:1,000, immunoprecipitation (IP): 2-5 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_006383.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... NOP56(10528)

Immunogen

The epitope recognized by PLA0266 maps to a region between residue 544 and 594 of human NOP56 ribonucleoprotein homolog using the numbering given in entry NP_006383.2 (GeneID 10528).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Nucleolar protein 56 (NOP56) is a core component of the box C/D snoRNP complex. The snoRNP core complex is made up of NOP58, NOP56, and fibrillin and is involved in ribosome biogenesis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA049918HPA021062PLA0256
description

Powered by Bethyl Laboratories, Inc.

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Powered by Bethyl Laboratories, Inc.

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

human

species reactivity

human

species reactivity

rat, mouse, human

species reactivity

human, mouse, rat

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

technique(s)

western blot: 1:2,000- 1:10,000, immunohistochemistry: 1:200- 1:1,000, immunoprecipitation (IP): 2-5 μg/mg

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:200-1:500, immunoblotting: 0.04-0.4 μg/mL

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:200-1:500

technique(s)

immunohistochemistry: 1:200- 1:1,000, proximity ligation assay: suitable, western blot: 1:2,000-1:10,000


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters