Skip to Content
Merck
All Photos(3)

Key Documents

PLA0259

Sigma-Aldrich

Rabbit anti-RuvBL2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

48 kDa TATA box-binding protein-interacting protein, 48 kDa TBP-interacting protein, 51 kDa erythrocyte cytosolic protein, 51-KD, CGI-46, ECP-51, ECP51, INO80 complex subunit J, INO80J, RVB2, Reptin 52, Reptin52, RuvB (E coli homolog)-like 2, RuvB-like 2 (E. coli), TAP54-beta, TIH2, TIP48, TIP49b, TIP60-associated protein 54-beta, erythrocyte cytosolic protein, repressing pontin 52, reptin, reptin52 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 11,200.00

CZK 11,200.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
CZK 11,200.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 11,200.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:500- 1:2,000
immunoprecipitation (IP): 5-15 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_006657.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... RuvBL2(10856)

General description

RuvB-like 2 (RUVBL2), a transcription factor, belongs to the AAA+ protein subfamily of conserved ATPases linked with diverse cellular functions. A distinctive domain II (DII) protrudes from one side of the hexamer and defines two separate faces of the ring in RUVBL2.

Immunogen

The epitope recognized by PLA0259 maps to a region between residue 1 and 50 of human RuvB-like 2 using the numbering given in entry NP_006657.1 (GeneID 10856).

Biochem/physiol Actions

RuvB-like 2 (RUVBL2) plays a regulatory role in the Fanconia anemia (FA) pathway, the assembly of the telomerase holoenzyme, and nonsense-mediated mRNA decay (NMD). It may play a role in tumorigenesis and cancer progression. Overexpression of the RUVBL2 protein is observed in hepatocellular carcinoma (HCC), renal cell carcinoma, colorectal cancer, gastric cancer, breast cancer, and salivary gland cancer. It can be used as a promising target for preventing and treating HCC.

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

RuvB-like 2 (RuvBL2) is the second human homologue of the bacterial RuvB gene. Bacterial RuvB protein is a DNA helicase essential for homologous recombination and DNA double-strand break repair. Functional analysis showed that this protein has both ATPase and DNA helicase activities [taken from NCBI Entrez Gene (GeneID: 9400)].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multilevel regulation of RUVBL2 expression predicts poor prognosis in hepatocellular carcinoma.
Yan, et al.
Cancer Cell International, 19, 249-249 (2020)
Andres López-Perrote et al.
eLife, 9 (2020-11-19)
Nonsense-mediated mRNA decay (NMD) is a surveillance pathway that degrades aberrant mRNAs and also regulates the expression of a wide range of physiological transcripts. RUVBL1 and RUVBL2 AAA-ATPases form an hetero-hexameric ring that is part of several macromolecular complexes such

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service