Skip to Content
Merck

Skip To

PLA0231

Rabbit anti-G3BP1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ATP-dependent DNA helicase VIII, DNA helicase VIII, G3BP, G3BP-1, GAP SH3 domain-binding protein 1, GAP binding protein, GTPase activating protein (SH3 domain) binding protein 1, HDH-VIII, Ras-GTPase-activating protein SH3-domain-binding protein, RasGAP-associated endoribonuclease G3BP, hDH VIII

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
1
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): 2-5 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_005745.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... G3BP1(10146)

Immunogen

The epitope recognized by PLA0231 maps to a region between residue 275 and 325 of human GTPase activating protein (SH3 domain) binding protein 1 using the numbering given in entry NP_005745.1 (GeneID 10146).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Ras-GAP SH3 domain binding protein (G3BP1) contains an RNA recognition motif (RRM) and an N-terminal nuclear transport factor 2-like (NTF2-like) domain. It was first identified in a screen for proteins that bind the SH3 domain of the Ras GTPase activating protein (RasGAP). It has been shown to function downstream of Ras and play a role in RNA metabolism, signal transduction, and proliferation. G3BP1 is primarily cytoplasmic but has also been localized to the nucleus. G3BP1 has been shown to be overexpressed in human cancers.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4300380SAB4300064G8046
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

species reactivity

mouse, human

species reactivity

human

species reactivity

human

species reactivity

human, rat (predicted), mouse (predicted)

UniProt accession no.

Q13283

UniProt accession no.

Q13283

UniProt accession no.

Q13283

UniProt accession no.

Q13283


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters