Skip to Content
Merck

Skip To

PLA0161

Rabbit anti-MOF/MYST1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

FLJ14040, K(lysine) acetyltransferase 8, Lysine acetyltransferase 8, MOF, MOZ, MYST histone acetyltransferase 1, MYST-1, MYST1, SAS2 and TIP60 protein 1, YBF2/SAS3, ZC2HC8, hMOF, histone acetyltransferase MYST1, ortholog of Drosophila males absent on the first (MOF), probable histone acetyltransferase MYST1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
12 300,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
1

12 300,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:500- 1:2,000, immunoprecipitation (IP): 2-5 μg/mg, western blot: 1:1,000- 1:10,000

accession no.

BAB13924.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... MOF/MYST1(84148)

Immunogen

The epitope recognized by PLA0161 maps to a region between residue 1 and 50 of human males absent on the first (MYST histone acetyltransferase 1) using the numbering given in entry BAB13924.1 (GeneID 84148).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

MOF is a member of the MYST family of histone acetyltransferases (HAT). MOF has been shown to be required for histone H4 lysine K16-specific acetylation. MOF interacts with ATM (ataxia-telangiectasia-mutated) and may participate in the activation of ATM in response to DNA damage.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AV38985SAB4503405HPA066324
description

Powered by Bethyl Laboratories, Inc.

description

affinity isolated antibody

description

affinity isolated antibody

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

species reactivity

human, mouse

species reactivity

human, guinea pig, mouse, rabbit, dog, bovine, horse, rat

species reactivity

mouse, rat, human

species reactivity

human

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

Gene Information

rabbit ... MOF/MYST1(84148)

Gene Information

human ... MYST4(23522)

Gene Information

human ... HAT1(8520)

Gene Information

human ... KAT8(84148)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters