Skip to Content
Merck

Skip To

PLA0156

Rabbit anti-PPP5C Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

PP-T, PP5, PPP5, PPT, catalytic subunit, protein phosphatase 5, protein phosphatase T

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
9 270,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
-

9 270,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:200-1:1,000, immunoprecipitation (IP): 2-5 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_006238.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... PPP5C(5536)

Immunogen

The epitope recognized by PLA0156 maps to a region between residue 1 and 50 of human Protein Phosphatase 5, Catalytic Subunit using the numbering given in entry NP_006238.1 (GeneID 5536).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

PPP5C is the protein phosphatase 5 catalytic unit that is a member of the large PPP family of serine/threonine protein phosphatases. PPP5C plays a role in mediating stress-activated signaling pathways and the DNA damage response. Unlike PP1, PP2A, and PPP4, which function as dimeric or trimeric enzymes, PPP5 functions as a monomer. PPP5 exists as a regulatory N-terminal tetratricopeptide repeat (TPR) domain fused to a C-terminal phosphatase catalytic domain. PPP5C has been demonstrated to associate with and regulate ATM and DNA-PK.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PLA0154HPA029065HPA043837
description

Powered by Bethyl Laboratories, Inc.

description

Powered by Bethyl Laboratories, Inc.

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

mouse, human

species reactivity

human, mouse

species reactivity

rat, human

species reactivity

human

Gene Information

rabbit ... PPP5C(5536)

Gene Information

rabbit ... PPP1CA(5499)

Gene Information

human ... PPP5C(5536)

Gene Information

human ... PPP4C(5531)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters