Skip to Content
Merck

Skip To

PLA0097

Rabbit anti-CBP Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CBP, KAT3A, RSTS, RTS, Rubinstein-Taybi syndrome

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
1


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

immunoprecipitation (IP): 1-4 μg/mg, western blot: 1:1,000- 1:25,000

accession no.

NP_004371.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... CBP(1387)

Immunogen

The epitope recognized by PLA0097 maps to a region between residues 975 and 1025 of human CREB Binding Protein using the numbering given in entry NP_004371.1 (GeneID 1387).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

CBP [cAMP response element-binding protein (CREB)-binding protein] interacts with and is a transcriptional co-activator of a multitude of transcriptional regulators that includes CREB. CBP is structurally and functionally similar to p300 but is encoded by an independent gene. CBP coactivation may be achieved through the regulation of chromatin structure due to its ability to catalyze the acetylation of histones. CBP has potential tumor suppressor properties and is important to integrating cell-signaling cues for proliferation, differentiation and apoptosis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PLA0029SAB2100480PLA0096
description

Powered by Bethyl Laboratories, Inc.

description

Powered by Bethyl Laboratories, Inc.

description

affinity isolated antibody

description

Powered by Bethyl Laboratories, Inc.

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

human

species reactivity

human, mouse

species reactivity

bovine, dog, mouse, horse, human, rabbit, rat, guinea pig

species reactivity

human

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

technique(s)

immunoprecipitation (IP): 1-4 μg/mg, western blot: 1:1,000- 1:25,000

technique(s)

immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:1,000- 1:10,000

technique(s)

immunohistochemistry: suitable, western blot: suitable

technique(s)

ChIP: 10 μg, immunohistochemistry: 1:1,000- 1:5,000, immunoprecipitation (IP): 1-5 μg/mg, western blot: 1:2,000-1:10,000


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters