Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Please contact Customer Service for Availability | 13 000,00 CZK |
About This Item
13 000,00 CZK
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human, mouse
technique(s)
western blot: 1:2,000-1:10,000
accession no.
NP_005648.1
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
rabbit ... 53BP1(7158)
Immunogen
Physical form
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description Powered by Bethyl Laboratories, Inc. | description Powered by Bethyl Laboratories, Inc. | description - | description - |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| conjugate unconjugated | conjugate - | conjugate unconjugated | conjugate - |
| clone polyclonal | clone - | clone polyclonal | clone polyclonal |
| antibody form affinity purified immunoglobulin | antibody form affinity purified immunoglobulin | antibody form affinity purified immunoglobulin | antibody form affinity isolated antibody |
| species reactivity human, mouse | species reactivity mouse, human | species reactivity human | species reactivity human |
| Gene Information rabbit ... 53BP1(7158) | Gene Information human ... TP53BP1(7158) | Gene Information rabbit ... p53(7157) | Gene Information human ... TP53BP1(7158) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



