About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human, mouse
technique(s)
immunohistochemistry: 1:1,000-1:5,000, immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:2,000-1:10,000
accession no.
NP_002936.1
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
rabbit ... RPA70(6117)
Immunogen
Physical form
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| species reactivity human, mouse | species reactivity mouse, human | species reactivity human | species reactivity human |
| conjugate unconjugated | conjugate - | conjugate unconjugated | conjugate unconjugated |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| clone polyclonal | clone - | clone polyclonal | clone polyclonal |
| antibody form affinity purified immunoglobulin | antibody form affinity purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| technique(s) immunohistochemistry: 1:1,000-1:5,000, western blot: 1:2,000-1:10,000, immunoprecipitation (IP): 2-10 μg/mg | technique(s) immunohistochemistry: 1:1,000- 1:5,000, immunoprecipitation (IP): 1- 4 μg/mg, western blot: 1:2,000- 1:10,000 | technique(s) immunohistochemistry: 1:20- 1:50 | technique(s) immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



