Skip to Content
Merck

Skip To

PLA0054

Goat anti-Chk1 Antibody, Affinity Purified

Synonym(s):

1, CHK1, CHK1 (checkpoint; S.pombe) homolog, CHK1 checkpoint homolog, CHK1 checkpoint homolog (S. pombe), Checkpoint, Chk1-S, S. pombe, cell cycle checkpoint kinase, checkpoint kinase-1, homolog of

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
10 300,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
4

10 300,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: 1:2,500- 1:15,000

accession no.

NP_001265.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

goat ... Chk1(1111)

Immunogen

The epitope recognized by PLA0054 maps to a region between residues 200 and 250 of human Checkpoint Kinase 1 using the numbering given in entry NP_001265.1 (GeneID 1111).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Chk1 functions as a checkpoint protein kinase that is critical to the regulation of the Cdc25a and Cdc25c phosphatases that promote S and G2/M phase progression, respectively. Chk1 kinase activity inhibits cell cycle progression in the presence of DNA damage by phosphorylating Cdc25a and Cdc25c and causing the degradation of Cdc25a and cytoplasmic sequestration of Cdc25c.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

77-92-9

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library