Skip to Content
Merck

PENP

Penicillin G procaine salt hydrate

~1000 U/mg

Synonym(s):

Benzylpenicillin procaine salt, Procaine penicilline G hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H18N2O4S · C13H20N2O2· xH2O
CAS Number:
Molecular Weight:
570.70 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

specific activity

~1000 U/mg

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

mode of action

cell wall synthesis | interferes

SMILES string

O.CCN(CC)CCOC(=O)c1ccc(N)cc1.CC2(C)S[C@@H]3[C@H](NC(=O)Cc4ccccc4)C(=O)N3[C@H]2C(O)=O

InChI

1S/C16H18N2O4S.C13H20N2O2.H2O/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;1-3-15(4-2)9-10-17-13(16)11-5-7-12(14)8-6-11;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);5-8H,3-4,9-10,14H2,1-2H3;1H2/t11-,12+,14-;;/m1../s1

InChI key

KZDCMKVLEYCGQX-UDPGNSCCSA-N

General description

Chemical structure: β-lactam

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Chicoine et al.
Journal of veterinary pharmacology and therapeutics, 32(3), 289-295 (2009-08-04)
This paper describes the pharmacokinetic profile of procaine penicillin G after intraperitoneal (IP) administration in eight lactating dairy cows. Procaine pencillin G (PPG, 21 000 IU/kg) was deposited into the abdominal cavity of each cow following an incision in the
Mehmet Doganay et al.
Journal of infection and public health, 3(3), 98-105 (2010-09-28)
Anthrax is still an endemic disease in some countries in the world and has become a re-emerging disease in western countries with recent intentional outbreak. The aim of this study was to review our clinical experience with cutaneous anthrax cases.
U Kayabas et al.
Chemotherapy, 58(1), 34-43 (2012-02-22)
Cutaneous anthrax (CA) is the most common clinical presentation in human anthrax, but the duration of antibiotic therapy in naturally occurring CA is controversial. The aim of this study was to compare the clinical outcomes of patients receiving antibiotic treatment
Sabine Mann et al.
Foodborne pathogens and disease, 8(8), 861-867 (2011-03-09)
The objective of this longitudinal controlled trial was to determine the effect of systemic treatment with ceftiofur on antimicrobial susceptibility of fecal Escherichia coli isolates in dairy cows. Cows with metritis or interdigital necrobacillosis requiring systemic antimicrobial treatment were sequentially
Carsten Krüger et al.
World journal of pediatrics : WJP, 6(2), 125-131 (2010-05-22)
With 700,000 to 1.5 million new cases annually, congenital syphilis remains a major infectious cause of morbidity and mortality in neonates, infants and children in resource-poor countries. We therefore analyzed the extent of congenital syphilis in the pediatric patient population

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service