Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C36H70O2
CAS Number:
Molecular Weight:
534.94
UNSPSC Code:
12352211
MDL number:
assay
≥99%
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCCCCCCOC(=O)CCCCCCC\C=C/CCCCCC
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
12 - Non Combustible Liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
