Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
30 ea | Please contact Customer Service for Availability | CZK 32,800.00 |
About This Item
CZK 32,800.00
material
black polystyrene , flat bottom clear polystyrene wells
sterility
sterile
feature
lid, polystyrene/polymer base
packaging
case of 30 ea (internal packs of 10)
manufacturer/tradename
Nunc 142761
maximum volume
500 μL
plate L × W
128 mm × 86 mm
size
384 wells
well maximum volume
100 μm
working volume
100 μL
suitability
suitable for (fluorescence/luminescence based assays)
General description
Optimum clarity for viewing well contents. Polymer bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications. Polystyrene upper structures available in black or white. Flat bottom well geometry for plate reader access. Black plates provide minimum back-scatterd light. White plates maximize reflection and provide minimum autoflorescence and autoluminescence. Note: All plates supplied with lid and sterile.
White or black upper structure with polystyrene/polymer bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications.
Application
HTS Applications.
Features and Benefits
- White or black upper structures with optically clear bottoms.
- Can be read from bottom and top, or both.
- Optical film made of polystyrene.
- Rounded square wells eliminate wicking (capillary action).
- Optimized for scintillation counting.
- Nunclon™ Δ (cell culture surface), Poly-D-Lysine or Collagen I surfaces for cell culture.
- Produced in clean environment.
- Working volume range: 10-100 μl/well.
- For research use only
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| material black polystyrene , flat bottom clear polystyrene wells | material flat bottom clear polystyrene wells, white polystyrene | material flat bottom white wells, polystyrene | material black polystyrene , flat bottom clear wells |
| manufacturer/tradename Nunc 142761 | manufacturer/tradename Nunc 142762 | manufacturer/tradename Nunc 165306 | manufacturer/tradename Nunc 165305 |
| feature lid, polystyrene/polymer base | feature lid, polystyrene/polymer base | feature lid, polystyrene/polymer base | feature lid, polystyrene/polymer base |
| working volume 100 μL | working volume 100 μL | working volume 200 μL | working volume 200 μL |
| well maximum volume 100 μm | well maximum volume 100 μm | well maximum volume 400 μm | well maximum volume - |
| suitability suitable for (fluorescence/luminescence based assays) | suitability suitable for (fluorescence/luminescence based assays) | suitability suitable for (fluorescence/luminescene based assays) | suitability suitable for (fluorescence/luminescene based assays) |
Still not finding the right product?
Explore all of our products under Nunc® MicroWell™ 384 well optical bottom plates
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



