Skip to Content
Merck

Skip To

P8404

Phenyltoloxamine citrate salt

Synonym(s):

N,N-Dimethyl-2([α-phenyl-o-tolyl]oxy)ethylamine

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C17H21NO · C6H8O7
CAS Number:
Molecular Weight:
447.48
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
214-644-7
MDL number:


form

powder

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CN(C)CCOc1ccccc1Cc2ccccc2

InChI

1S/C17H21NO.C6H8O7/c1-18(2)12-13-19-17-11-7-6-10-16(17)14-15-8-4-3-5-9-15;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-11H,12-14H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

ZZYHCCDMBJTROG-UHFFFAOYSA-N



Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters