Skip to Content
Merck
All Photos(1)

Key Documents

P7675

Sigma-Aldrich

Hexaethylene glycol monodecyl ether

Synonym(s):

C10E6, Decylhexaglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9(OCH2CH2)6OH
CAS Number:
Molecular Weight:
422.60
Beilstein:
1801568
MDL number:
UNSPSC Code:
12352200

description

non-ionic

density

0.987 g/mL at 25 °C (lit.)
0.99 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C22H46O7/c1-2-3-4-5-6-7-8-9-11-24-13-15-26-17-19-28-21-22-29-20-18-27-16-14-25-12-10-23/h23H,2-22H2,1H3

InChI key

GLGQRQQFWLTGES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service