Skip to Content
Merck
All Photos(1)

Key Documents

P7510

SAFC

5-Phenyl-1,10-phenanthroline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12N2
CAS Number:
Molecular Weight:
256.30
EC Number:
UNSPSC Code:
12352200

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Strahs et al.
Journal of molecular biology, 244(5), 494-510 (1994-12-16)
The cI repressor protein (cI) maintains bacteriophage lambda in the lysogenic state in infected Escherichia coli cells by binding cooperatively to three tandemly repeated sequences comprising the right operator (OR). Cooperative interactions occur between alternate pairs of cI dimers bound
A S Kimball et al.
Nucleic acids research, 23(15), 3009-3017 (1995-08-11)
The Flp protein catalyzes a site-specific recombination reaction between two 47 bp DNA sites without the assistance of any other protein or cofactor. The Flp recognition target (FRT) site consists of three nearly identical sequences, two of which are separated
E Hidalgo et al.
The Journal of biological chemistry, 270(36), 20908-20914 (1995-09-08)
SoxR protein of Escherichia coli is activated by superoxide-generating agents or nitric oxide as a powerful transcription activator of the soxS gene, whose product activates approximately 10 other promoters. SoxR contains non-heme iron essential for abortive initiation of transcription in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service