Sign In to View Organizational & Contract Pricing.
About This Item
NACRES:
NB.22
UNSPSC Code:
41122800
Manufacturer/tradename:
Bel-Art F18982-0009
Feature:
Holds 9 in. plates
Material:
polycarbonate (autoclavable)
Packaging:
pack of 1 ea
material
polycarbonate (autoclavable)
feature
Holds 9 in. plates
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18982-0009
H
24 cm (9.5 in.)
color
natural
compatibility
for use with Petri Dishes
General description
Increase workspace and efficiently transport large quantities of Petri dishes. The 24.1cm (9½") high size rack holds forty-eight 100mm Petri dishes, and the 36.8cm (14½") high rack holds eighty-four 100mm Petri dishes.
Application
Increase workspace and efficiently transport petri dishes with the Easyplate. The Easyplate eliminates a "balancing act" of petri dishes and enables you to transport petri dishes rapidly and orderly.
Features and Benefits
- Silicone holding strips grip each Petri dish; easily insert and remove groups of dishes without disturbing upper or lower dishes.
- Side labels provide space for critical sample identification.
- Lightweight, chemical resistant polycarbonate.
- Autoclavable at 121ºC (250ºF).
Legal Information
Scienceware is a registered trademark of Bel-Art Products, a division of SP
Still not finding the right product?
Explore all of our products under Scienceware® Easyplate petri dish transport carrier
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.