Skip to Content
Merck

Skip To

P5991

Nunc® 384 well polystyrene plates

384 well plate, non-treated, polystyrene, 120ul/well, pinchbar design, 30/cs

Synonym(s):

384 multiwell plate, 384 well microplate, 384 well microtiter plate, 384 well plate, Nunc 242757

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 case
Please contact Customer Service for Availability
CZK 11,400.00

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
flat bottom clear polystyrene wells, polystyrene
Size:
384 wells
Sterility:
sterile
Binding type:
non-treated surface
Feature:
lid

CZK 11,400.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom clear polystyrene wells, polystyrene

sterility

sterile

feature

lid

packaging

case of 30 ea (internal packs of 10)

manufacturer/tradename

Nunc 242757

maximum volume

120 μL

plate L × W

128 mm × 86 mm

size

384 wells

well maximum volume

120 μm

working volume

100 μL

binding type

non-treated surface

General description

Nunc® 384-well clear polystyrene plates with cell culture treated and non-treated surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Plates are high quality for consistent and reliable results.

Nunc™384-well polystyrene plates have rounded square-well geometry, eliminating wickin

Application

  • Immunoassays
  • Colorimetric assays and sample storage
  • Luminescence and fluorescence assays

Features and Benefits

  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Flexible working volumes; 10-100μL

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z722995Z723010TMO164688
binding type

non-treated surface

binding type

non-treated surface

binding type

non-treated surface

binding type

-

sterility

sterile

sterility

non-sterile

sterility

non-sterile

sterility

sterile

feature

lid

feature

lid: no

feature

lid: no

feature

lid

material

flat bottom clear polystyrene wells

material

black polystyrene , flat bottom black wells

material

clear polystyrene , flat bottom clear polystyrene wells

material

clear , polystyrene

manufacturer/tradename

Nunc 242757

manufacturer/tradename

Nunc 262260

manufacturer/tradename

Nunc 262160

manufacturer/tradename

Nunc 164688

size

384 wells

size

384 wells

size

384 wells

size

384 wells


Still not finding the right product?

Explore all of our products under Nunc® 384 well polystyrene plates




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library