Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
HS(CH2)3SH
CAS Number:
Molecular Weight:
108.23
EC Number:
203-706-9
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
1071197
vapor density
>1 (vs air)
vapor pressure
5 mmHg ( 37.7 °C)
refractive index
n20/D 1.539 (lit.)
bp
169 °C (lit.)
mp
−79 °C (lit.)
density
1.078 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
SCCCS
InChI
1S/C3H8S2/c4-2-1-3-5/h4-5H,1-3H2
InChI key
ZJLMKPKYJBQJNH-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Flam. Liq. 3 - Skin Irrit. 2
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
138.2 °F - closed cup
flash_point_c
59 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
109-80-8
CAS No.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

