Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C20H28O6
CAS Number:
Molecular Weight:
364.43
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
form
powder
color
white
storage temp.
−20°C
SMILES string
C[C@@H]1[C@@H](O)[C@]2(O)[C@H]([C@@H]3C=C(CO)C[C@]4(O)[C@@H](C=C(C)C4=O)[C@@]13O)C2(C)C
InChI
1S/C20H28O6/c1-9-5-13-18(24,15(9)22)7-11(8-21)6-12-14-17(3,4)20(14,26)16(23)10(2)19(12,13)25/h5-6,10,12-14,16,21,23-26H,7-8H2,1-4H3/t10-,12+,13-,14-,16-,18+,19-,20-/m1/s1
InChI key
QGVLYPPODPLXMB-FYYCTCHMSA-N
Application
Phorbol that is not biologically active; can be used as a negative control
signalword
Danger
Hazard Classifications
Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
