Skip to Content
Merck

P4855

Poly(ADP-ribose) Polymerase Automodified bovine

solution

Synonym(s):

PARP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
MDL number:
EC Number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Application

Positive control for immunoblotting techniques. A streak of M.W. > 116 kDa is detected by immunoblotting, representing PARP modified to various extents by poly(ADP-ribose). Recommended use: 5-20 μl per gel lane.

Other Notes

Partially purified bovine PARP, automodified by the addition of NAD and necessary cofactors.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library