Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Conjugate:
unconjugated
Clone:
DCS-100, monoclonal
Application:
ARR, ICC, IHC (p), IP, WB
Citations:
3
biological source
mouse
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
DCS-100, monoclonal
mol wt
antigen 19 kDa
contains
15 mM sodium azide
species reactivity
human
technique(s)
immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, immunoprecipitation (IP): suitable, microarray: suitable, western blot: 1:1,000 using a cultured human tumor cell line extract
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CDKN2D(1032)
General description
Cyclin dependent kinase inhibitor 2D (CDKN2D), also referred to as p19INK4d, is part of the inhibitors of cyclin-dependent kinases (INK4) family. It consists of 166 amino acids and five ankyrin-like repeats that aid in protein-protein interactions. The gene encoding this protein is localized on human chromosome 19p13.
Immunogen
recombinant human p19INK4d.
Application
Monoclonal Anti-p19INK4d antibody produced in mouse has been used in microarray.
Biochem/physiol Actions
Cyclin dependent kinase inhibitor 2D (CDKN2D) functions as a modulator of the cell cycle and has a role in DNA damage response. It acts as a chromatin accessibility factor. The protein associates with cyclin-dependent kinase 4 and 6 and inhibits their kinase activity.
Shows no cross-reactivity with other closely related Cdk inhibitors, such as p15INK4b, p16INK4a and p18INK4c.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.