Skip to Content
Merck
All Photos(1)

Key Documents

P3494

Sigma-Aldrich

Potassium clavulanate

from Streptomyces clavuligerus

Synonym(s):

Clavulanate potassium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8NO5K
CAS Number:
Molecular Weight:
237.25
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

biological source

Streptomyces clavuligerus

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[K+].[H][C@@]12CC(=O)N1[C@@H](C([O-])=O)C(\O2)=C\CO

InChI

1S/C8H9NO5.K/c10-2-1-4-7(8(12)13)9-5(11)3-6(9)14-4;/h1,6-7,10H,2-3H2,(H,12,13);/q;+1/p-1/b4-1-;/t6-,7-;/m1./s1

InChI key

ABVRVIZBZKUTMK-JSYANWSFSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

β-lactamase inhibitor, typically added to amoxicillin to increase its effectiveness.
In combination with penicillin group antibiotics to inhibit specific (group 2) β-penicillinases and cephalosporinases.

related product

Product No.
Description
Pricing

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Resp. Sens. 1 - Self-heat. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gustavo Jesus Vazquez-Zapien et al.
Romanian journal of morphology and embryology = Revue roumaine de morphologie et embryologie, 60(1), 189-194 (2019-07-03)
Kidney diseases are a global public health problem. Despite significant advances in the understanding of renal failure (RF) and replacement therapies, this condition carries a series of complications and the life's quality of patients decreases. Differentiation capability of stem cells
Winfried V Kern et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(9), 1149-1156 (2013-01-30)
This double-blind, multicenter trial compared the efficacy and safety of a single daily oral dose of moxifloxacin with oral combination therapy in low-risk febrile neutropenic patients with cancer. Inclusion criteria were cancer, febrile neutropenia, low risk of complications as predicted
Thomas Kwong et al.
Antimicrobial agents and chemotherapy, 56(9), 4845-4855 (2012-07-04)
Streptomyces clavuligerus produces a collection of five clavam metabolites, including the clinically important β-lactamase inhibitor clavulanic acid, as well as four structurally related metabolites called 5S clavams. The paralogue gene cluster of S. clavuligerus is one of three clusters of
Krisztina M Papp-Wallace et al.
Antimicrobial agents and chemotherapy, 56(8), 4428-4438 (2012-06-13)
β-Lactamases are important antibiotic resistance determinants expressed by bacteria. By studying the mechanistic properties of β-lactamases, we can identify opportunities to circumvent resistance through the design of novel inhibitors. Comparative amino acid sequence analysis of class A β-lactamases reveals that
Refaat B Hamed et al.
Natural product reports, 30(1), 21-107 (2012-11-09)
The β-lactam antibiotics and related β-lactamase inhibitors are amongst the most important small molecules in clinical use. Most, but not all, β-lactams including penicillins, cephalosporins, and clavulanic acid are produced via fermentation or via modification of fermented intermediates, with important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service