Skip to Content
Merck

P3144

D-Phe-Val-p-nitroanilide

protease substrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H24N4O4
CAS Number:
Molecular Weight:
384.43
UNSPSC Code:
12352204
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

storage temp.

−20°C

SMILES string

CC(C)C(NC(=O)C(N)Cc1ccccc1)C(=O)Nc2ccc(cc2)N(=O)=O

InChI

1S/C20H24N4O4/c1-13(2)18(20(26)22-15-8-10-16(11-9-15)24(27)28)23-19(25)17(21)12-14-6-4-3-5-7-14/h3-11,13,17-18H,12,21H2,1-2H3,(H,22,26)(H,23,25)

InChI key

MFFZQZIWTDHXEY-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library