Skip to Content
Merck

P259

1-O-Palmityl-sn-glycero-3-phosphocholine

powder

Synonym(s):

1-Hexadecyl-sn-glycero-3-phosphocholine, 3-sn-Lysophosphatidylcholine, 1-hexadecyl, Lyso-PAF-C16, Lyso-platelet activating factor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H52NO6P
CAS Number:
Molecular Weight:
481.65
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

color

white

solubility

H2O: 20 mg/mL

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COCCCCCCCCCCCCCCCC

InChI

1S/C24H52NO6P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-20-29-22-24(26)23-31-32(27,28)30-21-19-25(2,3)4/h24,26H,5-23H2,1-4H3/t24-/m1/s1

InChI key

VLBPIWYTPAXCFJ-XMMPIXPASA-N

Application

1-O-Palmityl-sn-glycero-3-phosphocholine also known as Lyso-PAF-(C16) can be used as a standard in the assay of platelet-activating factor.

Biochem/physiol Actions

Accumulation of lyso-PAF in atherogenic aortas may lead to unregulated PAF formation and can cause atherosclerosis.

Preparation Note

20mg of 1-O-Palmityl-sn-glycero-3-phosphocholine dissolves in 1 ml of water.

Other Notes

Inactive form of PAF[C16]

Disclaimer

Hygroscopic


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library