Skip to Content
Merck

P2482

Monoclonal Anti-Protein Kinase Bα antibody produced in mouse

clone PKB-175, ascites fluid

Synonym(s):

Anti-Akt1, Anti-PKBα

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44
MDL number:
Conjugate:
unconjugated
Clone:
PKB-175, monoclonal
Application:
ARR, ELISA (i), WB
Citations:
11
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

PKB-175, monoclonal

mol wt

antigen 56 kDa

contains

15 mM sodium azide

species reactivity

chicken, human, bovine, rat, mouse

technique(s)

indirect ELISA: suitable, microarray: suitable, western blot: 1:5,000 using human MCF-7 cell extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKT1(207)
mouse ... Akt1(11651)
rat ... Akt1(24185)

General description

Monoclonal Anti-Protein Kinase Bα (PKBα/Akt1) (mouse IgG1 isotype) is derived from the PKB-175 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from a BALB/c mouse. PKB/Akt is composed of an N-terminal pleckstrin-homology (PH) domain, followed by a catalytic kinase domain and a short C-terminal regulatory domain. This cytoplasmic protein is made up of 480 amino acids. AKT1 is mapped to human chromosome 14q32.
Protein Kinase B (also known as Akt, or RAC-PK) is a protein that belongs to serine/threonine kinases.

Immunogen

Synthetic peptide corresponding to amino acids 461-477 of human PKBα/Akt1, conjugated to KLH. The sequence is identical in mouse, rat, and bovine PKBα/Akt1, highly conserved in PKBβ/Akt2, and diverges in PKBγ.

Application

Monoclonal Anti-Protein Kinase Bα antibody produced in mouse has been used in:
  • western blot
  • immunofluorescence
  • enzyme-linked immunosorbent assay (ELISA)

Biochem/physiol Actions

Monoclonal Anti-Protein Kinase Bα (PKBα/Akt1) recognizes PKB isoform α.
Protein kinase Bα (PKBα/Akt1) plays a crucial role as a suppressor of apoptotic cell death in different cell types, induced by a variety of stimuli including growth factor withdrawal, loss of cell adhesion and DNA damage. It can protect cerebellar neurons from apoptosis induced by insulin-like growth factor 1 (IGF-1) withdrawal. Akt is the cellular homologue of the viral oncogene v-akt of the AKT-8 acute transforming retrovirus found in rodent T cell lymphoma. It is considered to play an important role in the control of cell cycle, cell proliferation and differentiation . AKT plays a key role in angiogenesis.

Physical form

Monoclonal Anti-Protein Kinase Bα (PKBα/Akt1) is supplied as ascites fluid containing 15 mM sodium azide

Preparation Note

For continuous use, store at 2-8C for upto one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library