Skip to Content
Merck

P2178

Pro-Asp-Val-Asp-His-Val-Phe-Leu-Arg-Phe-NH2

≥97% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C59H86N16O14
CAS Number:
Molecular Weight:
1243.41
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](NC(=O)[C@H](Cc2c[nH]cn2)NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H]3CCCN3)C(C)C)C(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc4ccccc4)C(N)=O

Biochem/physiol Actions

Cardioinhibitory; inhibits contraction of the oviduct visceral muscle, but potentiates contraction of the extensortibiae muscle

Other Notes

FMRF-amide-like neuropeptide originally isolated from locust (Schistocera gregaria)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library