Skip to Content
Merck

Skip To

P1300

Pluronic® F-68

solid, BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

Polyoxyethylene-polyoxypropylene block copolymer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(C3H6O.C2H4O)x
CAS Number:
UNSPSC Code:
12352207
PubChem Substance ID:
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Contains 100 ppm BHT, non-ionic

product line

BioReagent

form

solid

mol wt

average mol wt 8350

technique(s)

cell culture | insect: suitable, cell culture | mammalian: suitable

CMC

0.04 mM (20-25°C)

HLB

29

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

Application

A non-ionic detergent that protects cells from hydrodynamic damage.

Legal Information

Pluronic is a registered trademark of BASF


Storage Class

11 - Combustible Solids

WGK

WGK 1

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–5 of 5 Questions  
  1. What is the difference between Pluronic® F-68 Products P1300 and P7061?

    1 answer
    1. The main differences between the P1300 and the P7061 are the testing specifications, and the form in which they are provided.  The P1300 is more of a white prill/solid, and the P7061 is more of a powder.

      Helpful?

  2. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

  3. Will the P130Pluronic® F-68, Product P1300, meet NF, Clinical or Pharamaceutical specifications or not?

    1 answer
    1. The P1300 is not an NF, Clinical nor Pharamaceutical grade product; it will not necessarily meet those specifications.

      Helpful?

  4. Can Pluronic® F-68, Product P1300, be sterilized by filtration?

    1 answer
    1. Solutions of Product No. P1300 can be filtered by using a 0.2 mm pore size filter.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters