Skip to Content
Merck

P0958

1,2-Dioctanoyl-sn-glycero-3-phosphocholine

~99%

Synonym(s):

L-α-Phosphatidylcholine, dioctanoyl, Dioctanoyl-L-α-phosphatidylcholine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H48NO8P
CAS Number:
Molecular Weight:
509.61
UNSPSC Code:
12352211
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~99%

storage temp.

−20°C

SMILES string

CCCCCCCC(=O)OC[C@@H](COP(O)(=O)OCC[N](C)(C)C)OC(=O)CCCCCCC



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library