Skip to Content

P0865

N-(3-Pyridyl)indomethacinamide

≥98%

Synonym(s):

N-(3-Pyridyl)-1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetamide, N-3PyIA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H20N3O3Cl
CAS Number:
Molecular Weight:
433.89
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

form

powder

solubility

chloroform: 10 mg/mL to clear, colorless to light yellow

shipped in

wet ice

storage temp.

−20°C

SMILES string

COc1ccc2n(c(C)c(CC(=O)Nc3cccnc3)c2c1)C(=O)c4ccc(Cl)cc4

InChI

1S/C24H20ClN3O3/c1-15-20(13-23(29)27-18-4-3-11-26-14-18)21-12-19(31-2)9-10-22(21)28(15)24(30)16-5-7-17(25)8-6-16/h3-12,14H,13H2,1-2H3,(H,27,29)

InChI key

DTAPHQYAZUTCIG-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective reversible cyclooxygenase-2 (COX-2) inhibitor (IC50 = 0.1 μM, human recombinant); about 300-fold less potent as an inhibitor of human recombinant COX-1. Not as selective with natural ovine COX (IC50 = 75 μM for COX-1 and 50 μM for COX-2).
Potent and selective reversible cyclooxygenase-2 (COX-2) inhibitor. Not as selective with natural ovine COX.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library