mol wt
5,000-15,000
storage temp.
−20°C
Analysis Note
Molecular weight based on viscosity.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.