OH30428204
Ohaus® Compass Portable Balance
Model CR221, weighing capacity 220 g, resolution: 0.1 g
Synonym(s):
laboratory balance, movable balance, transportable balance, travel balance, weighing balance
About This Item
Recommended Products
material
plastic weighing plate
feature
CE compliant
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30428204
weighing capacity measuring range
220 g, resolution: 0.1 g
weighing pan W × W
13.2 cm × 12.5 cm
battery power
alkaline, 1000 hr life
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service