Skip to Content
Merck

O9762

N-Oleoylglycine

≥98%

Synonym(s):

N-(1-Oxo-9-octadecenyl)-(Z)-glycine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H37NO3
CAS Number:
Molecular Weight:
339.51
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352211
EC Number:
220-009-5
MDL number:
eCl@ss:
32160703
Assay:
≥98%
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

form

solid

lipid type

neutral glycerides

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCCC/C=C\CCCCCCCC(NCC(O)=O)=O

InChI

1S/C20H37NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19(22)21-18-20(23)24/h9-10H,2-8,11-18H2,1H3,(H,21,22)(H,23,24)/b10-9-

InChI key

HPFXACZRFJDURI-KTKRTIGZSA-N

Biochem/physiol Actions

N-Oleoylglycine is equally efficient as oleamide in reducing locomotion and hypothermia.
Enzymatically produced in mammals. Substrate for peptidylglycine α-amidating enzyme.

Preparation Note

Prepare stock solutions (10 mg/mL) by dissolving the crystalline solid in an organic solvent such as ethanol, methanol, acetone, acetonitrile, or DMSO. Working dilutions may be made in aqueous buffers.


Still not finding the right product?

Explore all of our products under N-Oleoylglycine


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library