Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.47
UNSPSC Code:
12171500
biological source
synthetic
density
>1.20 g/mL
application(s)
hematology
histology
storage temp.
room temp
Application
For use with Osteo-Bed Bone Embedding Kit. Solvent for removal of plastic from sections prior to rehydration and staining. Most histochemical and immunohistochemistry staining protocols require the removal of methyl methacrylate (MMA) and rehydration of mounted sections for improved stain penetration and greater contrast.
signalword
Danger
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
75-09-2
CAS No.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


